Anti-Monocyte + Macrophage Rat Monoclonal Antibody [clone: MOMA-2]

Lieferant: Abcam
ABCAAB33451-125EA 941 EUR
ABCAAB33451-125
Anti-Monocyte + Macrophage Rat Monoclonal Antibody [clone: MOMA-2]
Antikörper
Type: Primary
Antigen: Monocyte + Macrophage
Clonality: monoclonal
Clone: MOMA-2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR