Anti-IL10 Rat Monoclonal Antibody [clone: JES5-2A5]

Lieferant: Abcam
ABCAAB33471-100EA 812 EUR
ABCAAB33471-100
Anti-IL10 Rat Monoclonal Antibody [clone: JES5-2A5]
Antikörper
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: JES5-2A5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR