Anti-IRF5 Mouse Monoclonal Antibody [clone: 10T1]

Lieferant: Abcam
ABCAAB33478-100EA 719 EUR
ABCAAB33478-100
Anti-IRF5 Mouse Monoclonal Antibody [clone: 10T1]
Antikörper
Type: Primary
Antigen: IRF5 (interferon regulatory factor 5)
Clonality: monoclonal
Clone: 10T1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR