Anti-ANPEP Rat Monoclonal Antibody [clone: R3-63]

Lieferant: Abcam
ABCAAB33489-100EA 877 EUR
ABCAAB33489-100
Anti-ANPEP Rat Monoclonal Antibody [clone: R3-63]
Antikörper
Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: monoclonal
Clone: R3-63
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR