Anti-THBD Mouse Monoclonal Antibody [clone: 0.N.186]

Lieferant: Abcam
ABCAAB33513-250EA 666 EUR
ABCAAB33513-250
Anti-THBD Mouse Monoclonal Antibody [clone: 0.N.186]
Antikörper
Type: Primary
Antigen: THBD (thrombomodulin)
Clonality: monoclonal
Clone: 0.N.186
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR