Anti-ATIC Mouse monoclonal antibody unconjugated [clone: F38 P7 H9]

Lieferant: Abcam
ABCAAB33520-50EA 688 EUR
ABCAAB33520-50
Anti-ATIC Mouse monoclonal antibody unconjugated [clone: F38 P7 H9]
Antikörper
Type: Primary
Antigen: ATIC (5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase)
Clonality: monoclonal
Clone: F38 P7 H9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR