Anti-SSR1 Mouse Monoclonal Antibody [clone: 0.T.59]

Lieferant: Abcam
ABCAAB33552-100EA 690 EUR
ABCAAB33552-100
Anti-SSR1 Mouse Monoclonal Antibody [clone: 0.T.59]
Antikörper
Type: Primary
Antigen: SSR1 (signal sequence receptor, alpha)
Clonality: monoclonal
Clone: 0.T.59
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR