Anti-CYP1B1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB33586-100EA 703 EUR
ABCAAB33586-100
Anti-CYP1B1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP1B1 (cytochrome P450, family 1, subfamily B, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR