Anti-TNNT2 Mouse Monoclonal Antibody [clone: 0.N.590]

Lieferant: Abcam
ABCAAB33589-100EA 682 EUR
ABCAAB33589-100
Anti-TNNT2 Mouse Monoclonal Antibody [clone: 0.N.590]
Antikörper
Type: Primary
Antigen: TNNT2 (troponin T type 2 (cardiac))
Clonality: monoclonal
Clone: 0.N.590
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR