Anti-Prostate Specific Antigen Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB33609-250EA 666 EUR
ABCAAB33609-250
Anti-Prostate Specific Antigen Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Prostate Specific Antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR