Anti-Hemoglobin A1c Sheep Polyclonal Antibody

Lieferant: Abcam
ABCAAB33615-500EA 666 EUR
ABCAAB33615-500
Anti-Hemoglobin A1c Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Hemoglobin A1c
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR