Anti-TSHB Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB33632-500EA 690 EUR
ABCAAB33632-500
Anti-TSHB Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TSHB (thyroid stimulating hormone, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR