Anti-NTRK2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB33655-100EA 728 EUR
ABCAAB33655-100
Anti-NTRK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NTRK2 (neurotrophic tyrosine kinase, receptor, type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR