Anti-ITGA2B Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MWReg30]

Lieferant: Abcam
ABCAAB33660-50EA 428 EUR
ABCAAB33660-50
Anti-ITGA2B Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MWReg30]
Antikörper
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone: MWReg30
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR