Anti-CD81 Armenian Hamster Monoclonal Antibody [clone: Eat 2]

Lieferant: Abcam
ABCAAB33697-125EA 696 EUR
ABCAAB33697-125
Anti-CD81 Armenian Hamster Monoclonal Antibody [clone: Eat 2]
Antikörper
Type: Primary
Antigen: CD81 (CD81 molecule)
Clonality: monoclonal
Clone: Eat 2
Conjugation: unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG1
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR