Anti-SLAMF1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9D1]

Lieferant: Abcam
ABCAAB33708-50EA 322 EUR
ABCAAB33708-50
Anti-SLAMF1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9D1]
Antikörper
Type: Primary
Antigen: SLAMF1 (signaling lymphocytic activation molecule family member 1)
Clonality: monoclonal
Clone: 9D1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR