Anti-CD200R1 Rat Monoclonal Antibody [clone: OX-110]

Lieferant: Abcam
ABCAAB33736-100EA 666 EUR
ABCAAB33736-100
Anti-CD200R1 Rat Monoclonal Antibody [clone: OX-110]
Antikörper
Type: Primary
Antigen: CD200R1 (CD200 receptor 1)
Clonality: monoclonal
Clone: OX-110
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR