Anti-CD200 / OX2 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MRC OX90]
Lieferant: Abcam
ABCAAB33737-50EA 480 EUR
ABCAAB33737-50
Anti-CD200 / OX2 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MRC OX90]
Antikörper
Type: Primary
Antigen: CD200 / OX2
Clonality: monoclonal
Clone: MRC OX90
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse