Anti-IL10RA Rat Monoclonal Antibody [clone: 1B1.3a]

Lieferant: Abcam
ABCAAB33738-50EA 666 EUR
ABCAAB33738-50
Anti-IL10RA Rat Monoclonal Antibody [clone: 1B1.3a]
Antikörper
Type: Primary
Antigen: IL10RA (interleukin 10 receptor, alpha)
Clonality: monoclonal
Clone: 1B1.3a
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR