Anti-CD200 / OX2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MRC OX90]

Lieferant: Abcam
ABCAAB33739-50EA 360 EUR
ABCAAB33739-50
Anti-CD200 / OX2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MRC OX90]
Antikörper
Type: Primary
Antigen: CD200 / OX2
Clonality: monoclonal
Clone: MRC OX90
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR