Anti-EIF4E Rabbit monoclonal antibody unconjugated [clone: Y449]

Lieferant: Abcam
ABCAAB33768-100EA 688 EUR
ABCAAB33768-100
Anti-EIF4E Rabbit monoclonal antibody unconjugated [clone: Y449]
Antikörper
Type: Primary
Antigen: EIF4E (eukaryotic translation initiation factor 4E)
Clonality: monoclonal
Clone: Y449
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR