Anti-SHC1 Rabbit Monoclonal Antibody [clone: Y332]
Lieferant: Abcam
ABCAAB33770-100EA 897 EUR
ABCAAB33770-100
Anti-SHC1 Rabbit Monoclonal Antibody [clone: Y332]
Antikörper
Type: Primary
Antigen: SHC1 (SHC (Src homology 2 domain containing) transforming protein 1)
Clonality: monoclonal
Clone: Y332
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen