Anti-IL1B Rabbit Monoclonal Antibody [clone: EP408Y]

Lieferant: Abcam
ABCAAB33774-100EA 672 EUR
ABCAAB33774-100
Anti-IL1B Rabbit Monoclonal Antibody [clone: EP408Y]
Antikörper
Type: Primary
Antigen: IL1B (interleukin 1, beta)
Clonality: monoclonal
Clone: EP408Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR