Anti-CD4 Mouse Monoclonal Antibody [clone: OX-35]
Lieferant: Abcam
ABCAAB33775-100EA 882 EUR
ABCAAB33775-100
Anti-CD4 Mouse Monoclonal Antibody [clone: OX-35]
Antikörper
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: OX-35
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat