Anti-SLC2A4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB33780-100EA 697 EUR
ABCAAB33780-100
Anti-SLC2A4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC2A4 (solute carrier family 2 (facilitated glucose transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR