Anti-PECAM1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: TLD-3A12]

Lieferant: Abcam
ABCAAB33858-50EA 328 EUR
ABCAAB33858-50
Anti-PECAM1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: TLD-3A12]
Antikörper
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: TLD-3A12
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR