Anti-CD22 Rabbit Monoclonal Antibody [clone: EP498Y]

Lieferant: Abcam
ABCAAB33859-100EA 697 EUR
ABCAAB33859-100
Anti-CD22 Rabbit Monoclonal Antibody [clone: EP498Y]
Antikörper
Type: Primary
Antigen: CD22 (CD22 molecule)
Clonality: monoclonal
Clone: EP498Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR