Anti-MAP2K6 Rabbit Monoclonal Antibody [clone: EP557Y]

Lieferant: Abcam
ABCAAB33866-100EA 732 EUR
ABCAAB33866-100
Anti-MAP2K6 Rabbit Monoclonal Antibody [clone: EP557Y]
Antikörper
Type: Primary
Antigen: MAP2K6 (mitogen-activated protein kinase kinase 6)
Clonality: monoclonal
Clone: EP557Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR