Anti-SPN Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: W3/13]

Lieferant: Abcam
ABCAAB33885-50EA 375 EUR
ABCAAB33885-50
Anti-SPN Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: W3/13]
Antikörper
Type: Primary
Antigen: SPN (sialophorin)
Clonality: monoclonal
Clone: W3/13
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR