Anti-UB Rabbit Monoclonal Antibody [clone: EP296YT]

Lieferant: Abcam
ABCAAB33893-100EA 697 EUR
ABCAAB33893-100
Anti-UB Rabbit Monoclonal Antibody [clone: EP296YT]
Antikörper
Type: Primary
Antigen: UB (ubiquitin)
Clonality: monoclonal
Clone: EP296YT
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR