Anti-ICAM1 Mouse Monoclonal Antibody [clone: 1A29]

Lieferant: Abcam
ABCAAB33894-50EA 672 EUR
ABCAAB33894-50
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 1A29]
Antikörper
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone: 1A29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR