Anti-BRAF Rabbit monoclonal antibody unconjugated [clone: EP152Y]

Lieferant: Abcam
ABCAAB33899-100EA 981 EUR
ABCAAB33899-100
Anti-BRAF Rabbit monoclonal antibody unconjugated [clone: EP152Y]
Antikörper
Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: monoclonal
Clone: EP152Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR