Anti-BCL6 Rabbit monoclonal antibody unconjugated [clone: EP529Y]

Lieferant: Abcam
ABCAAB33901-100EA 743 EUR
ABCAAB33901-100
Anti-BCL6 Rabbit monoclonal antibody unconjugated [clone: EP529Y]
Antikörper
Type: Primary
Antigen: BCL6 (B-cell CLL/lymphoma 6)
Clonality: monoclonal
Clone: EP529Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR