Anti-SMAD1 Rabbit Monoclonal Antibody [clone: EP565Y]

Lieferant: Abcam
ABCAAB33902-100EA 713 EUR
ABCAAB33902-100
Anti-SMAD1 Rabbit Monoclonal Antibody [clone: EP565Y]
Antikörper
Type: Primary
Antigen: SMAD1 (SMAD family member 1)
Clonality: monoclonal
Clone: EP565Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR