Anti-BBC3 Rabbit monoclonal antibody unconjugated [clone: EP512Y]

Lieferant: Abcam
ABCAAB33906-100EA 762 EUR
ABCAAB33906-100
Anti-BBC3 Rabbit monoclonal antibody unconjugated [clone: EP512Y]
Antikörper
Type: Primary
Antigen: BBC3 (BCL2 binding component 3)
Clonality: monoclonal
Clone: EP512Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR