Anti-MAP2K4 Rabbit Monoclonal Antibody [clone: EP615Y]

Lieferant: Abcam
ABCAAB33912-100EA 749 EUR
ABCAAB33912-100
Anti-MAP2K4 Rabbit Monoclonal Antibody [clone: EP615Y]
Antikörper
Type: Primary
Antigen: MAP2K4 (mitogen-activated protein kinase kinase 4)
Clonality: monoclonal
Clone: EP615Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR