Anti-LYN Rabbit Monoclonal Antibody [clone: EP504Y]

Lieferant: Abcam
ABCAAB33914-100EA 688 EUR
ABCAAB33914-100
Anti-LYN Rabbit Monoclonal Antibody [clone: EP504Y]
Antikörper
Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: monoclonal
Clone: EP504Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR