Anti-TRAF6 Rabbit Monoclonal Antibody [clone: EP591Y]
Lieferant: Abcam
ABCAAB33915-100EA 1070 EUR
ABCAAB33915-100
Anti-TRAF6 Rabbit Monoclonal Antibody [clone: EP591Y]
Antikörper
Type: Primary
Antigen: TRAF6 (TNF receptor-associated factor 6, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: EP591Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen