Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MRC OX-1]

Lieferant: Abcam
ABCAAB33916-50EA 423 EUR
ABCAAB33916-50
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MRC OX-1]
Antikörper
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: MRC OX-1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR