Anti-MALT1 Rabbit Monoclonal Antibody [clone: EP603Y]

Lieferant: Abcam
ABCAAB33921-100EA 917 EUR
ABCAAB33921-100
Anti-MALT1 Rabbit Monoclonal Antibody [clone: EP603Y]
Antikörper
Type: Primary
Antigen: MALT1 (mucosa associated lymphoid tissue lymphoma translocation gene 1)
Clonality: monoclonal
Clone: EP603Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR