Anti-GFAP Rabbit monoclonal antibody unconjugated [clone: EP672Y]

Lieferant: Abcam
ABCAAB33922-100EA 999 EUR
ABCAAB33922-100
Anti-GFAP Rabbit monoclonal antibody unconjugated [clone: EP672Y]
Antikörper
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: monoclonal
Clone: EP672Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR