Anti-TFRC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: OX-26]

Lieferant: Abcam
ABCAAB33927-50EA 666 EUR
ABCAAB33927-50
Anti-TFRC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: OX-26]
Antikörper
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: monoclonal
Clone: OX-26
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR