Anti-CD45RC Mouse Monoclonal Antibody [clone: MRC OX-22]

Lieferant: Abcam
ABCAAB33945-100EA 666 EUR
ABCAAB33945-100
Anti-CD45RC Mouse Monoclonal Antibody [clone: MRC OX-22]
Antikörper
Type: Primary
Antigen: CD45RC
Clonality: monoclonal
Clone: MRC OX-22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR