Anti-Sendai virus Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB33988-1EA 666 EUR
ABCAAB33988-1
Anti-Sendai virus Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Sendai virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR