Anti-TNFRSF4 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: OX-86]

Lieferant: Abcam
ABCAAB33998-50EA 328 EUR
ABCAAB33998-50
Anti-TNFRSF4 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: OX-86]
Antikörper
Type: Primary
Antigen: TNFRSF4 (tumor necrosis factor receptor superfamily, member 4)
Clonality: monoclonal
Clone: OX-86
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR