Anti-Ovine MHC Class II DR Mouse Monoclonal Antibody [clone: 37.68]

Lieferant: Abcam
ABCAAB34007-125EA 657 EUR
ABCAAB34007-125
Anti-Ovine MHC Class II DR Mouse Monoclonal Antibody [clone: 37.68]
Antikörper
Type: Primary
Antigen: Ovine MHC Class II DR
Clonality: monoclonal
Clone: 37.68
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR