Anti-PDGF AB + BB Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB34074-250EA 690 EUR
ABCAAB34074-250
Anti-PDGF AB + BB Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDGF AB + BB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR