Anti-SIRP alpha Mouse Monoclonal Antibody [clone: ED9]

Lieferant: Abcam
ABCAAB34086-125EA 877 EUR
ABCAAB34086-125
Anti-SIRP alpha Mouse Monoclonal Antibody [clone: ED9]
Antikörper
Type: Primary
Antigen: SIRP alpha
Clonality: monoclonal
Clone: ED9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR