Anti-IL8 Mouse Monoclonal Antibody [clone: 8M6]

Lieferant: Abcam
ABCAAB34100-125EA 672 EUR
ABCAAB34100-125
Anti-IL8 Mouse Monoclonal Antibody [clone: 8M6]
Antikörper
Type: Primary
Antigen: IL8 (interleukin 8)
Clonality: monoclonal
Clone: 8M6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR