Anti-CD200R1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-102]

Lieferant: Abcam
ABCAAB34135-50EA 666 EUR
ABCAAB34135-50
Anti-CD200R1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-102]
Antikörper
Type: Primary
Antigen: CD200R1 (CD200 receptor 1)
Clonality: monoclonal
Clone: OX-102
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR